1-Cyclopropyl-1-phenylethanol 95%

Supplier: Ambeed
Warning

Synonyms: α-Cyclopropyl-α-methylbenzenemethanol

A193973-1G A193973-250MG
103816-604EA 185 USD
103816-604 103847-212 77085-120 77236-988
1-Cyclopropyl-1-phenylethanol 95%
1-Cyclopropyl-1-phenylethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Boiling Pt: 241…242 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00019243
CAS Number: 5558-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR