Cyclobutylhydrazine hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: 1-Cyclobutylhydrazine hydrochloride

A678296-250MG A678296-25G
103840-530EA 141 USD
103840-530 77018-518 77891-336
Cyclobutylhydrazine hydrochloride 95%
Cyclobutylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂
Flash Pt: 80.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06658415
CAS Number: 158001-21-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR