1-Chloropyrrolo[1,2-a]pyrazine 98%

Supplier: Ambeed
Warning

A187527-100MG A187527-250MG A187527-1G A187527-5G A187527-25G
103816-020EA 64 USD
103816-020 103846-610 76813-804 76813-806 77084-304 77084-306 77883-399
1-Chloropyrrolo[1,2-a]pyrazine 98%
1-Chloropyrrolo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12400782
CAS Number: 136927-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR