1-Chloro-2-naphthol 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloronaphthalen-2-ol

A130558-100G A130558-10G A130558-250MG A130558-1G A130558-25G A130558-5G
103801-442EA 9 USD
103801-442 77234-500 77234-502 77234-504 77313-888 77313-890 77313-892
1-Chloro-2-naphthol 97%
1-Chloro-2-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇ClO
MW: 178.62 g/mol
Boiling Pt: 306.5 °C (760 mm Hg)
Melting Pt: 70 °C
Flash Pt: 139.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00997084
CAS Number: 633-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR