1-Chloroethyl (4-nitrophenyl) carbonate 97%

Supplier: Ambeed
Warning

A163644-250MG A163644-1G A163644-10G
103805-740EA 6 USD
103805-740 103844-000 77163-288 77320-104 77611-594
1-Chloroethyl (4-nitrophenyl) carbonate 97%
1-Chloroethyl (4-nitrophenyl) carbonate

Precaución: Sealed in dry, at 2…8°C

Formula: C₉H₈ClNO₅
Boiling Pt: 359.1±42.0 °C (760 mm Hg)
Melting Pt: 71.0…71.7 °C
Flash Pt: 171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19441847
CAS Number: 101623-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR