1-Chlorocarbonyl-4-piperidinopiperidine hydrochloride 97%

Supplier: Ambeed
Danger

A120028-100MG A120028-10G A120028-250MG A120028-25G A120028-1G A120028-5G
103799-870EA 9 USD
103799-870 76995-748 76995-750 76995-752 76995-754 77311-978 77311-980
1-Chlorocarbonyl-4-piperidinopiperidine hydrochloride 97%
1-Chlorocarbonyl-4-piperidinopiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₀Cl₂N₂O
MW: 267.2 g/mol
Flash Pt: 162.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08445613
CAS Number: 143254-82-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR