1-Chloro-4-propoxy-9H-thioxanthen-9-one 97%

Supplier: Ambeed
Warning

A356296-1G A356296-5G
103826-166EA 12 USD
103826-166 103837-174 77306-680 77306-682
1-Chloro-4-propoxy-9H-thioxanthen-9-one 97%
1-Chloro-4-propoxy-9H-thioxanthen-9-one

Precaución: for Research Use Only

Formula: C₁₆H₁₃ClO₂S
MW: 304.79 g/mol
Boiling Pt: 467.4 °C (760 mm Hg)
Melting Pt: 100.0…103.0 °C
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00191928
CAS Number: 142770-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR