1-Chloro-4-iodobenzene 98%

Supplier: Ambeed
Warning

A155568-1000G A155568-100G A155568-500G A155568-25G A155568-5G
103842-976EA 5 USD
103842-976 77110-468 77110-470 77110-472 77110-474 77300-128 77300-130
1-Chloro-4-iodobenzene 98%
1-Chloro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₄ClI
Boiling Pt: 227 °C (760 mm Hg)
Melting Pt: 53…54 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001053
CAS Number: 637-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR