1-Chloro-3-fluoro-2-iodobenzene 97%

Supplier: Ambeed
Warning

A151735-10G A151735-1G A151735-25G
103804-296EA 23 USD
103804-296 103842-446 76973-138 77318-030 77318-032
1-Chloro-3-fluoro-2-iodobenzene 97%
1-Chloro-3-fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFI
Storage Temperature: Refrigerator
MDL Number: MFCD08458092
CAS Number: 127654-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR