1-Chloro-3-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethoxy)chlorobenzene

A729640-5G A729640-10G
103840-780EA 15 USD
103840-780 76845-008 77281-024
1-Chloro-3-(trifluoromethoxy)benzene 98%
1-Chloro-3-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
MW: 196.55 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00276971
CAS Number: 772-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR