1-Chloro-2-methylpropyl propionate 95%

Supplier: Ambeed
Warning

A175591-1G A175591-5G A175591-10G A175591-25G
103807-062EA 25 USD
103807-062 103845-374 77082-714 77082-716 77236-404 77236-406
1-Chloro-2-methylpropyl propionate 95%
1-Chloro-2-methylpropyl propionate

Precaución: for Research Use Only

Formula: C₇H₁₃ClO₂
MW: 164.63 g/mol
Boiling Pt: 166.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09033184
CAS Number: 58304-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR