1-Chloro-2-iodobenzene 98% stabilized

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-1-iodobenzene

Stabilization: Stabilized with Copper chip

A223951-5G A223951-100G A223951-10G A223951-1G A223951-500G A223951-25G
103849-960EA 6 USD
103849-960 77088-592 77237-692 77237-694 77237-696 77237-698 77303-680
1-Chloro-2-iodobenzene 98% stabilized
1-Chloro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 234…235 °C (760 mm Hg)
Melting Pt: 1 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001033
CAS Number: 615-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR