1-Chloro-2-fluoro-4-methoxy-5-nitrobenzene 97%
Supplier: Ambeed
|
Warning
|
A133987-10G
A133987-250MG
A133987-1G
A133987-5G
103801-946EA
46
USD
103801-946
76972-256
76972-258
77314-646
77314-648
1-Chloro-2-fluoro-4-methoxy-5-nitrobenzene 97%
1-Chloro-2-fluoro-4-methoxy-5-nitrobenzene
Precaución: for Research Use Only
Formula:
C₇H₅ClFNO₃ Boiling Pt: 273.3 °C (760 mm Hg) Flash Pt: 119.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD11847613 CAS Number: 1089280-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...