4-Chloro-3-fluorobenzotrifluoride 95%
Supplier: Ambeed
|
Warning
|
A536246-1G
A536246-25G
A536246-5G
103828-594EA
43
USD
103828-594
103839-714
76843-736
76843-738
76843-740
4-Chloro-3-fluorobenzotrifluoride 95%
4-Chloro-3-fluorobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₇H₃ClF₄ MW: 198.55 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00060646 CAS Number: 32137-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...