1-Chloro-2-fluoro-3-iodobenzene 95%

Supplier: Ambeed
Warning

A203739-1G A203739-25G A203739-5G A203739-100G A203739-10G A203739-500G
103817-486EA 5 USD
103817-486 103848-146 77086-278 77086-280 77086-282 77152-246 77152-248 77152-250
1-Chloro-2-fluoro-3-iodobenzene 95%
1-Chloro-2-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFI
MW: 256.4439 g/mol
Boiling Pt: 229.9 °C (760 mm Hg)
Flash Pt: 92.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094152
CAS Number: 72373-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR