(2-Chlorophenyl)acetylene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-ethynylbenzene

A131428-1G A131428-25G A131428-5G
103801-576EA 13 USD
103801-576 77298-480 77314-078 77314-080
(2-Chlorophenyl)acetylene 97%
(2-Chlorophenyl)acetylene

Precaución: for Research Use Only

Formula: C₈H₅Cl
MW: 136.58 g/mol
Boiling Pt: 82 °C (15 mm Hg)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269951
CAS Number: 873-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR