1-Chloro-2-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Danger

A228785-25G A228785-5G A228785-100G A228785-10G A228785-500G
103850-326EA 5 USD
103850-326 77089-132 77089-134 77237-820 77237-822 77237-824
1-Chloro-2-(trifluoromethoxy)benzene 98%
1-Chloro-2-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
Boiling Pt: 143.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03840206
CAS Number: 450-96-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR