1-Cbz-piperazine 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl piperazine-1-carboxylate, Piperazine-1-carboxylic Acid Benzyl Ester, 1-piperazinecarboxylic acid, phenylmethyl ester, Benzyl 1-piperazinecarboxylate

A196076-100G A196076-10G A196076-1G A196076-25G A196076-5G
103816-808EA 5 USD
103816-808 103847-422 77164-418 77164-420 77302-494 77302-496 77302-498
1-Cbz-piperazine 97%
1-Cbz-piperazine

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₁₂H₁₆N₂O₂
Boiling Pt: 352.4 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00274317
CAS Number: 31166-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR