N-(Benzyloxycarbonyl)azetidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Cbz-Azetidine-3-carboxylic acid

A292801-1G A292801-5G A292801-100G A292801-100MG
103813-954EA 8 USD
103813-954 103835-092 76837-692 76837-694 77154-052 77154-054
N-(Benzyloxycarbonyl)azetidine-3-carboxylic acid 97%
N-(Benzyloxycarbonyl)azetidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MW: 235.239 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027504
CAS Number: 97628-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR