1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Benzyl 3-hydroxyazetidine-1-carboxylate, 1-Cbz-3-Hydroxyazetidine

A162071-10G A162071-1G A162071-25G A162071-5G A162071-100G
103805-566EA 9 USD
103805-566 103843-814 77011-232 77319-842 77319-844 77319-846 77611-578
1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester 97%
1-Azetidinecarboxylic acid-3-hydroxyphenylmethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
Boiling Pt: 356.7 °C (760 mm Hg)
Flash Pt: 169.518 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09037849
CAS Number: 128117-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR