1-Butylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Butylboronic acid, n-Butylboronic acid
A108096-10G
A108096-25G
A108096-1G
A108096-5G
103797-296EA
6
USD
103797-296
103823-070
76809-796
76809-798
77309-916
77309-918
1-Butylboronic acid 98%
1-Butylboronic acid
Precaución: for Research Use Only
Formula:
C₄H₁₁BO₂ MW: 101.94 g/mol Boiling Pt: 189.2 °C (760 mm Hg) Melting Pt: 94…96 °C Flash Pt: 68.2 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002106 CAS Number: 4426-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...