1-Bromo-2-naphthol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromonaphthalen-2-ol, 1-Bromo-2-hydroxynaphthalene
A639350-100G
A639350-10G
A639350-1G
A639350-500G
A639350-25G
A639350-5G
103840-332EA
8
USD
103840-332
76806-298
76806-300
76806-302
76806-304
76844-510
77308-040
1-Bromo-2-naphthol 98%
1-Bromo-2-naphthol
Precaución: for Research Use Only
Formula:
C₁₀H₇BrO MW: 223.07 g/mol Boiling Pt: 313.7 °C (760 mm Hg) Melting Pt: 78…81 °C Flash Pt: 143.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003869 CAS Number: 573-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...