1-Bromoisoquinoline 97%
Supplier: Ambeed
|
Danger
|
A112876-100MG
A112876-250MG
A112876-25G
A112876-5G
103798-124EA
9
USD
103798-124
103823-960
76687-650
76687-652
76687-654
77310-790
1-Bromoisoquinoline 97%
1-Bromoisoquinoline
Precaución: for Research Use Only
Formula:
C₉H₆BrN MW: 208.0546 g/mol Boiling Pt: 316.3 °C (760 mm Hg) Melting Pt: 43…47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00234478 CAS Number: 1532-71-4 UN: 2923 ADR: 8,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...