1-Bromoheptan-2-one 95%

Supplier: Ambeed
Warning

A265102-250MG A265102-1G
103812-706EA 347 USD
103812-706 103852-600 76988-236 77304-910
1-Bromoheptan-2-one 95%
1-Bromoheptan-2-one

Precaución: for Research Use Only

Formula: C₇H₁₃BrO
MW: 193.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00233012
CAS Number: 16339-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR