1-Bromoethyl acetate 95%

Supplier: Ambeed
Warning

A720661-100G A720661-5G
103840-740EA 15 USD
103840-740 77199-006 77308-202
1-Bromoethyl acetate 95%
1-Bromoethyl acetate

Precaución: for Research Use Only

Formula: C₄H₇BrO₂
MW: 167.001 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08061392
CAS Number: 40258-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR