1-Bromocarbazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-9H-carbazole

A157915-100G A157915-100MG A157915-10G A157915-250MG A157915-500G A157915-1G A157915-25G A157915-5G
103805-060EA 17 USD
103805-060 103843-266 76871-930 76871-932 76871-934 76871-936 76871-938 77319-102 77319-104 77319-106
1-Bromocarbazole 97%
1-Bromocarbazole

Precaución: for Research Use Only

Formula: C₁₂H₈BrN
Boiling Pt: 409.2±18.0 °C (760 mm Hg)
Melting Pt: 111…112 °C
Flash Pt: 201.3 °C
Storage Temperature: Ambient
MDL Number: MFCD18450164
CAS Number: 16807-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR