1-Bromo-5-nitronaphthalene 97%
Supplier: Ambeed
|
Warning
|
A109828-100G
A109828-100MG
A109828-10G
A109828-250MG
A109828-25G
A109828-1G
A109828-5G
103797-570EA
8
USD
103797-570
103823-374
77093-944
77093-946
77093-948
77093-950
77093-952
77283-888
77310-182
1-Bromo-5-nitronaphthalene 97%
1-Bromo-5-nitronaphthalene
Precaución: for Research Use Only
Formula:
C₁₀H₆BrNO₂ Boiling Pt: 363.8 °C (760 mm Hg) Flash Pt: 173.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156397 CAS Number: 5328-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...