4-Bromo-2-fluoro-5-nitroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-5-fluoro-4-methoxy-2-nitrobenzene

A101413-250MG A101413-1G A101413-5G A101413-10G
103795-962EA 11 USD
103795-962 103821-646 76869-152 77282-726 77297-096 77610-746
4-Bromo-2-fluoro-5-nitroanisole 97%
4-Bromo-2-fluoro-5-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₃
Boiling Pt: 288 °C (760 mm Hg)
Melting Pt: 95…97 °C
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD11100974
CAS Number: 661463-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR