2-Bromo-4-fluoro-6-nitrotoluene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-5-fluoro-2-methyl-3-nitrobenzene

A766285-100G A766285-10G A766285-250MG A766285-5G A766285-1G A766285-25G
103840-912EA 13 USD
103840-912 76791-444 76791-446 76791-448 76845-144 77308-286 77308-288
2-Bromo-4-fluoro-6-nitrotoluene 97%
2-Bromo-4-fluoro-6-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MW: 234.0225 g/mol
Boiling Pt: 259.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03094189
CAS Number: 502496-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR