1-Bromo-5-chloropentane 95%

Supplier: Ambeed
Warning

A127256-100G A127256-500G A127256-25G
103808-354EA 120 USD
103808-354 77234-362 77234-364 77234-366 77313-186
1-Bromo-5-chloropentane 95%
1-Bromo-5-chloropentane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₃CH₂Cl
Boiling Pt: 211 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00001016
CAS Number: 54512-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR