1-Bromo-4-propylbenzene 98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)propane, 1-Bromo-4-n-propylbenzene

A145570-100G A145570-10G A145570-25G A145570-5G
103841-644EA 6 USD
103841-644 76689-160 76689-162 77316-952 77316-954
1-Bromo-4-propylbenzene 98%
1-Bromo-4-propylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 224.3 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00012456
CAS Number: 588-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR