1-Bromo-4-phenylbutane 96%

Supplier: Ambeed
Warning

A906363-1G A906363-25G A906363-5G A906363-100G
103841-360EA 50 USD
103841-360 76845-628 76845-630 76845-632 77118-800
1-Bromo-4-phenylbutane 96%
1-Bromo-4-phenylbutane

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br
Boiling Pt: 54…56 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00154988
CAS Number: 13633-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR