1-Bromo-4-octylbenzene 95%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)octane

A289474-1G A289474-25G A289474-5G A289474-100G A289474-10G
103813-832EA 8 USD
103813-832 103834-972 76837-546 76837-548 76837-550 77099-298 77099-300
1-Bromo-4-octylbenzene 95%
1-Bromo-4-octylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₂₁Br
MW: 269.22 g/mol
Boiling Pt: 313.7 °C (760 mm Hg)
Flash Pt: 176.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00038348
CAS Number: 51554-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR