1-Bromo-4-methylnaphthalene 96%

Supplier: Ambeed
Warning

A527679-100G A527679-10G A527679-1G A527679-25G A527679-5G
103828-546EA 7 USD
103828-546 103839-658 76805-250 76805-252 76843-672 76843-674 76843-676
1-Bromo-4-methylnaphthalene 96%
1-Bromo-4-methylnaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 303.7 °C (760 mm Hg)
Melting Pt: 5.5…6.0 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003872
CAS Number: 6627-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR