4-Bromo-3-methylanisole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene
A188395-100G
A188395-10G
A188395-1G
A188395-500G
A188395-25G
A188395-5G
103846-688EA
6
USD
103846-688
76998-904
76998-906
76998-908
76998-910
77084-414
77084-416
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole
Precaución: for Research Use Only
Formula:
C₈H₉BrO MW: 201.06 g/mol Boiling Pt: 235.3 °C (760 mm Hg) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060687 CAS Number: 27060-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...