1-Bromo-4-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-iodobenzene

A147399-100G A147399-10G A147399-500G A147399-1000G A147399-25G
103808-694EA 197 USD
103808-694 103841-870 77286-002 77286-004 77286-006 77299-578 77317-242
1-Bromo-4-iodobenzene 98%
1-Bromo-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 244.8 °C (760 mm Hg)
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00001051
CAS Number: 589-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR