2-Bromo-5-iodotoluene 98%

Supplier: Ambeed
Warning

A185438-100G A185438-10G A185438-500G A185438-25G A185438-5G
103846-420EA 8 USD
103846-420 76769-712 76769-714 76769-716 77084-058 77301-974
2-Bromo-5-iodotoluene 98%
2-Bromo-5-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrI
Boiling Pt: 264.2 °C (760 mm Hg)
Flash Pt: 113.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00070749
CAS Number: 202865-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR