2-Bromo-5-iodoanisole 97%

Supplier: Ambeed
Warning

A288966-25G A288966-5G A288966-100G A288966-10G A288966-1G
103813-814EA 5 USD
103813-814 103834-956 76837-524 76837-526 77262-794 77262-796 77305-548
2-Bromo-5-iodoanisole 97%
2-Bromo-5-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
Boiling Pt: 293.7 °C (760 mm Hg)
Flash Pt: 131.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07780651
CAS Number: 755027-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR