2-Bromo-5-iodobenzotrifluoride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-4-iodo-2-(trifluoromethyl)benzene, 2-Bromo-5-iodo-α,α,α-trifluorotoluene
A169433-100G
A169433-1G
A169433-25G
A169433-5G
103806-384EA
6
USD
103806-384
103844-640
76813-170
77081-716
77081-718
77081-720
2-Bromo-5-iodobenzotrifluoride 97%
2-Bromo-5-iodobenzotrifluoride
Precaución: for Research Use Only
Formula:
C₇H₃BrF₃I MW: 350.902 g/mol Melting Pt: 78…80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11855999 CAS Number: 364-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...