1-Bromo-4-hexylbenzene 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromophenyl)hexane

A116587-100G A116587-25G A116587-1G A116587-10G
103798-760EA 6 USD
103798-760 103824-624 77233-936 77233-938 77311-428 77566-112
1-Bromo-4-hexylbenzene 97%
1-Bromo-4-hexylbenzene

Precaución: For research use only.

Formula: C₁₂H₁₇Br
MW: 241.17 g/mol
Boiling Pt: 280 °C (760 mm Hg)
Flash Pt: 144.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00061114
CAS Number: 23703-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR