1-Bromo-4-fluoro-5-isopropyl-2-methoxybenzene 98%

Supplier: Ambeed
Warning

A135113-10G A135113-1G A135113-5G
103802-092EA 53 USD
103802-092 77149-792 77298-752 77298-754
1-Bromo-4-fluoro-5-isopropyl-2-methoxybenzene 98%
1-Bromo-4-fluoro-5-isopropyl-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
Boiling Pt: 240.1±40.0 °C (760 mm Hg)
Flash Pt: 124.3 °C
Storage Temperature: Ambient
MDL Number: MFCD15528104
CAS Number: 944317-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR