2-Bromo-5-fluorotoluene 98%

Supplier: Ambeed
Danger

A563621-10G A563621-25G A563621-100G
103839-900EA 5 USD
103839-900 76774-946 76844-012 76844-014 77878-359
2-Bromo-5-fluorotoluene 98%
2-Bromo-5-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrF
Boiling Pt: 181.4 °C (760 mm Hg)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00017921
CAS Number: 452-63-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR