2-Bromo-5-fluoroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-fluoro-2-methoxybenzene

A206906-100G A206906-1G A206906-5G A206906-10G
103817-744EA 9 USD
103817-744 103848-424 76770-354 77086-690 77086-692 77626-046
2-Bromo-5-fluoroanisole 97%
2-Bromo-5-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 187.9 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD04973752
CAS Number: 450-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR