1-Bromo-4-fluoro-2-methoxy-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A275336-1G A275336-25G A275336-5G A275336-10G A275336-250MG
103813-244EA 29 USD
103813-244 103853-170 77093-018 77093-020 77093-022 77153-704 77153-706
1-Bromo-4-fluoro-2-methoxy-5-nitrobenzene 95%
1-Bromo-4-fluoro-2-methoxy-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₃
MW: 250.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25969224
CAS Number: 935288-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR