1-Bromo-4-fluoro-2-iodobenzene 98%

Supplier: Ambeed
Warning

A286732-25G A286732-5G A286732-100G A286732-1G A286732-10G
103813-726EA 9 USD
103813-726 103834-862 76837-404 76837-406 77153-916 77305-502 77685-816
1-Bromo-4-fluoro-2-iodobenzene 98%
1-Bromo-4-fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 241.8±20.0 °C (760 mm Hg)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00042578
CAS Number: 202865-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR