2-Bromo-α,α,α,5-tetrafluorotoluene 98%

Supplier: Ambeed
Warning

A218007-100G A218007-10G A218007-500G A218007-1000G A218007-25G A218007-5G
103809-530EA 265 USD
103809-530 103849-414 76770-746 76770-748 76770-750 77088-066 77088-068 77303-474
2-Bromo-α,α,α,5-tetrafluorotoluene 98%
2-Bromo-α,α,α,5-tetrafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
Boiling Pt: 160.4 °C (760 mm Hg)
Flash Pt: 63.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00040937
CAS Number: 40161-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR