1-Bromo-4-ethynylbenzene 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Bromophenylacetylene, 1-Ethynyl-4-bromobenzene, (4-Bromophenyl)acetylene, (4-Bromophenyl)ethyne
A200423-1G
A200423-25G
A200423-5G
A200423-10G
A200423-250MG
A200423-100G
103817-200EA
15
USD
103817-200
103847-828
77085-886
77085-888
77085-890
77192-266
77192-268
77615-702
1-Bromo-4-ethynylbenzene 97%
1-Bromo-4-ethynylbenzene
Precaución: for Research Use Only
Formula:
C₈H₅Br MW: 181.03 g/mol Melting Pt: 64…67 °C Storage Temperature: Freezer |
MDL Number:
MFCD00168822 CAS Number: 766-96-1 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...