1-Bromo-4-chloro-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A195884-25G A195884-5G A195884-10G
103847-404EA 5 USD
103847-404 77085-350 77085-352 77237-034
1-Bromo-4-chloro-2-nitrobenzene 97%
1-Bromo-4-chloro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
Boiling Pt: 242.5 °C (760 mm Hg)
Melting Pt: 68…72 °C
Flash Pt: 100.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00024320
CAS Number: 41513-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR