1-Bromo-4-chloro-2-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-chloroiodobenzene, 1-Bromo-4-Chloro-2-Iodobenzene

A256819-1G A256819-25G A256819-5G A256819-10G A256819-250MG
103812-286EA 11 USD
103812-286 103852-150 77091-652 77091-654 77091-656 77238-402 77238-404
1-Bromo-4-chloro-2-iodobenzene 98%
1-Bromo-4-chloro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrCl
MW: 317.3495 g/mol
Boiling Pt: 279 °C (760 mm Hg)
Melting Pt: 32…33 °C
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672943
CAS Number: 148836-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR