1-Bromo-4-(trans-4-pentylcyclohexyl)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-4-(trans-4-N-pentylcyclohexyl)benzene
A179415-100G
A179415-10G
103845-770EA
6
USD
103845-770
77287-114
77287-116
1-Bromo-4-(trans-4-pentylcyclohexyl)benzene 98%
1-Bromo-4-(trans-4-pentylcyclohexyl)benzene
Precaución: for Research Use Only
Formula:
C₁₇H₂₅Br MW: 309.2800 g/mol Boiling Pt: 363 °C (760 mm Hg) Flash Pt: 165.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00577359 CAS Number: 79832-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...